(5-Chloro-4-fluoro-2-methoxyphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

108672-1G 108672-500MG 108672-5G 108672-25G
77463-928EA 428.48 USD
77463-928 77476-732 77486-788 77497-630
(5-Chloro-4-fluoro-2-methoxyphenyl)(methyl)sulfane ≥95%
(5-Chloro-4-fluoro-2-methoxyphenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₈H₈ClFOS
MW: 206.665 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34779954
CAS Number: 2708092-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR