4-Bromo-2-(difluoromethoxy)-1-iodobenzene ≥95%

Supplier: AOBChem USA

Warning

108358-1G 108358-500MG 108358-5G 108358-25G
77463-924EA 211.15 USD
77463-924 77476-730 77486-782 77497-624
4-Bromo-2-(difluoromethoxy)-1-iodobenzene ≥95%
4-Bromo-2-(difluoromethoxy)-1-iodobenzene

Precaución: For research use only

Formula: C₇H₄BrF₂IO
MW: 348.911 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09878216
CAS Number: 1000575-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR