2-(4-Bromo-2-fluorophenyl)-2-methyl-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

10795-1G 10795-500MG 10795-5G 10795-25G
77463-920EA 139.05 USD
77463-920 77476-726 77486-778 77497-622
2-(4-Bromo-2-fluorophenyl)-2-methyl-1,3-dioxolane ≥97%
2-(4-Bromo-2-fluorophenyl)-2-methyl-1,3-dioxolane

Precaución: For research use only

Formula: C₁₀H₁₀BrFO₂
MW: 261.088 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09839107
CAS Number: 625446-23-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR