4-Bromo-2-fluoro-3-(trifluoromethyl)benzoic acid ≥95%

Supplier: AOBChem USA

Warning

107238-1G 107238-500MG 107238-5G 107238-25G
77463-904EA 325.48 USD
77463-904 77476-710 77486-762 77497-604
4-Bromo-2-fluoro-3-(trifluoromethyl)benzoic acid ≥95%
4-Bromo-2-fluoro-3-(trifluoromethyl)benzoic acid

Precaución: For research use only

Formula: C₈H₃BrF₄O₂
MW: 287.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22052785
CAS Number: 194804-96-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR