4-formyl-2-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

107207-1G 107207-500MG 107207-5G
77463-900EA 820.91 USD
77463-900 77476-706 77486-758
4-formyl-2-(trifluoromethyl)benzoic acid ≥97%
4-formyl-2-(trifluoromethyl)benzoic acid

Precaución: For research use only

Formula: C₉H₅F₃O₃
Storage Temperature: Ambient
MDL Number: MFCD28357239
CAS Number: 1289087-72-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR