3-Bromo-5-chloro-4-iodopyridine ≥97%

Supplier: AOBChem USA

Warning

107029-1G 107029-500MG 107029-5G 107029-25G
77463-896EA 177.16 USD
77463-896 77476-702 77486-754 77497-600
3-Bromo-5-chloro-4-iodopyridine ≥97%
3-Bromo-5-chloro-4-iodopyridine

Precaución: For research use only

Formula: C₅H₂BrClIN
Storage Temperature: Ambient
MDL Number: MFCD28128014
CAS Number: 1070870-39-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR