2,4-Dibromo-3-chloro-6-methylaniline ≥95%

Supplier: AOBChem USA

Warning

106301-1G 106301-500MG 106301-5G 106301-25G
77463-892EA 173.04 USD
77463-892 77476-698 77486-750 77497-590
2,4-Dibromo-3-chloro-6-methylaniline ≥95%
2,4-Dibromo-3-chloro-6-methylaniline

Precaución: For research use only

Formula: C₇H₆Br₂ClN
Storage Temperature: Ambient
MDL Number: MFCD09800824
CAS Number: 89794-16-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR