4-Bromo-5-hydroxy-2-methylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

106128-1G 106128-500MG 106128-5G
77463-888EA 685.98 USD
77463-888 77476-694 77486-746
4-Bromo-5-hydroxy-2-methylbenzoic acid ≥95%
4-Bromo-5-hydroxy-2-methylbenzoic acid

Precaución: For research use only

Formula: C₈H₇BrO₃
MW: 231.043 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34778888
CAS Number: 1781204-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR