3-Bromo-5-chloro-2-(4-morpholino)pyridine ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 4-(3-Bromo-5-chloropyridin-2-yl)morpholine

105259-1G 105259-5G 105259-25G
77463-880EA 64.89 USD
77463-880 77486-736 77497-558
3-Bromo-5-chloro-2-(4-morpholino)pyridine ≥95%
3-Bromo-5-chloro-2-(4-morpholino)pyridine

Precaución: For research use only

Formula: C₉H₁₀BrClN₂O
MW: 277.546 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13195782
CAS Number: 1199773-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR