≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Hydroxy-3,5-diisopropylbenzaldehyde

105101-1G 105101-500MG 105101-5G 105101-25G
77463-876EA 86.52 USD
77463-876 77476-684 77486-732 77497-548
≥97%
4-Hydroxy-3,5-bis(isopropyl)benzaldehyde

Precaución: For research use only

Formula: C₁₃H₁₈O₂
Storage Temperature: Ambient
MDL Number: MFCD00812875
CAS Number: 10537-86-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR