3,5-Diisopropyl-4-methoxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

105063-1G 105063-500MG 105063-5G
77463-872EA 676.71 USD
77463-872 77476-680 77486-728
3,5-Diisopropyl-4-methoxybenzaldehyde ≥95%
3,5-Diisopropyl-4-methoxybenzaldehyde

Precaución: For research use only

Formula: C₁₄H₂₀O₂
Storage Temperature: Ambient
MDL Number: MFCD34778302
CAS Number: 221445-27-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR