3-Bromo-5-fluoro-4-(methylthio)pyridine ≥97%

Supplier: AOBChem USA

Warning

104690-1G 104690-500MG 104690-5G
77463-868EA 497.49 USD
77463-868 77476-676 77486-724
3-Bromo-5-fluoro-4-(methylthio)pyridine ≥97%
3-Bromo-5-fluoro-4-(methylthio)pyridine

Precaución: For research use only

Formula: C₆H₅BrFNS
MW: 222.078 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34778159
CAS Number: 2901083-16-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR