2-Methoxy-5-propylbenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

104547-1G 104547-500MG 104547-5G 104547-25G
77463-864EA 134.93 USD
77463-864 77476-672 77486-720 77497-520
2-Methoxy-5-propylbenzaldehyde ≥95%
2-Methoxy-5-propylbenzaldehyde

Precaución: For research use only

Formula: C₁₁H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD06248102
CAS Number: 145742-54-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR