1-(5-Chloro-2-fluoro-4-methylphenyl)cyclohexanol ≥97%

Supplier: AOBChem USA

Warning

104263-1G 104263-500MG 104263-5G 104263-25G
77463-856EA 415.09 USD
77463-856 77476-664 77486-712 77497-510
1-(5-Chloro-2-fluoro-4-methylphenyl)cyclohexanol ≥97%
1-(5-Chloro-2-fluoro-4-methylphenyl)cyclohexanol

Precaución: For research use only

Formula: C₁₃H₁₆ClFO
Storage Temperature: Ambient
MDL Number: MFCD34778021
CAS Number: 2918866-63-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR