1-(2-Fluoro-4-methylphenyl)-4,4-dimethylcyclohexanol ≥95%

Supplier: AOBChem USA

Warning

104099-1G 104099-500MG 104099-5G 104099-25G
77463-852EA 415.09 USD
77463-852 77476-660 77486-708 77497-506
1-(2-Fluoro-4-methylphenyl)-4,4-dimethylcyclohexanol ≥95%
1-(2-Fluoro-4-methylphenyl)-4,4-dimethylcyclohexanol

Precaución: For research use only

Formula: C₁₅H₂₁FO
MW: 236.325 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34777871
CAS Number: 2918855-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR