2,2-Dibromo-1-(3-chloro-2-fluorophenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

104043-1G 104043-500MG 104043-5G 104043-25G
77463-848EA 81.37 USD
77463-848 77476-656 77486-704 77497-504
2,2-Dibromo-1-(3-chloro-2-fluorophenyl)ethanone ≥95%
2,2-Dibromo-1-(3-chloro-2-fluorophenyl)ethanone

Precaución: For research use only

Formula: C₈H₄Br₂ClFO
MW: 330.376 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34681240
CAS Number: 2901066-46-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR