5-(3-Bromothiophen-2-yl)-2-phenylpyrimidine ≥95%

Supplier: AOBChem USA

Warning

104031-1G 104031-500MG 104031-5G 104031-25G
77463-844EA 415.09 USD
77463-844 77476-652 77486-700 77497-500
5-(3-Bromothiophen-2-yl)-2-phenylpyrimidine ≥95%
5-(3-Bromothiophen-2-yl)-2-phenylpyrimidine

Precaución: For research use only

Formula: C₁₄H₉BrN₂S
Storage Temperature: Ambient
MDL Number: MFCD34777843
CAS Number: 2901103-22-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR