1-(2-Fluoro-5-methylphenyl)cyclopentanol ≥97%

Supplier: AOBChem USA

Warning

103415-1G 103415-500MG 103415-5G 103415-25G
77463-840EA 415.09 USD
77463-840 77476-648 77486-696 77497-490
1-(2-Fluoro-5-methylphenyl)cyclopentanol ≥97%
1-(2-Fluoro-5-methylphenyl)cyclopentanol

Precaución: For research use only

Formula: C₁₂H₁₅FO
MW: 194.245 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17222256
CAS Number: 1341302-84-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR