2,6-Difluoro-3-(1-hydroxycyclopentyl)benzonitrile ≥95%

Supplier: AOBChem USA

Warning

103385-1G 103385-500MG 103385-5G 103385-25G
77463-836EA 415.09 USD
77463-836 77476-644 77486-692 77497-488
2,6-Difluoro-3-(1-hydroxycyclopentyl)benzonitrile ≥95%
2,6-Difluoro-3-(1-hydroxycyclopentyl)benzonitrile

Precaución: For research use only

Formula: C₁₂H₁₁F₂NO
Storage Temperature: Ambient
MDL Number: MFCD34777566
CAS Number: 2918871-96-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR