1-(4-Bromo-3-chloro-2-fluorophenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

103375-1G 103375-500MG 103375-5G
77463-832EA 810.61 USD
77463-832 77476-640 77486-688
1-(4-Bromo-3-chloro-2-fluorophenyl)cyclobutanol ≥95%
1-(4-Bromo-3-chloro-2-fluorophenyl)cyclobutanol

Precaución: For research use only

Formula: C₁₀H₉BrClFO
MW: 279.533 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29498514
CAS Number: 1702577-77-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR