1-(Benzyloxy)-3,4-dibromo-2-fluorobenzene ≥95%

Supplier: AOBChem USA

Warning

103299-1G 103299-500MG 103299-5G 103299-25G
77463-828EA 262.65 USD
77463-828 77476-636 77486-684 77497-482
1-(Benzyloxy)-3,4-dibromo-2-fluorobenzene ≥95%
1-(Benzyloxy)-3,4-dibromo-2-fluorobenzene

Precaución: For research use only

Formula: C₁₃H₉Br₂FO
MW: 360.016 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29043949
CAS Number: 1881295-70-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR