1-(2-Fluoro-3-methylphenyl)-4,4-dimethylcyclohexanol ≥95%

Supplier: AOBChem USA

Warning

102694-1G 102694-5G 102694-500MG 102694-25G
77463-816EA 415.09 USD
77463-816 77486-672 77476-624 77497-446
1-(2-Fluoro-3-methylphenyl)-4,4-dimethylcyclohexanol ≥95%
1-(2-Fluoro-3-methylphenyl)-4,4-dimethylcyclohexanol

Precaución: For research use only

Formula: C₁₅H₂₁FO
MW: 236.325 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34777273
CAS Number: 2918929-30-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR