1-(5-Chlorothiophen-2-yl)-4,4-dimethylcyclohexanol ≥97%

Supplier: AOBChem USA

Warning

102690-1G 102690-5G 102690-500MG
77463-812EA 597.4 USD
77463-812 77486-668 77476-620
1-(5-Chlorothiophen-2-yl)-4,4-dimethylcyclohexanol ≥97%
1-(5-Chlorothiophen-2-yl)-4,4-dimethylcyclohexanol

Precaución: For research use only

Formula: C₁₂H₁₇ClOS
MW: 244.781 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23865105
CAS Number: 1511327-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR