1-(3,5-Dichloro-2-fluorophenyl)-4,4-dimethylcyclohexanol ≥95%

Supplier: AOBChem USA

Warning

102639-1G 102639-5G 102639-500MG 102639-25G
77463-808EA 415.09 USD
77463-808 77486-664 77476-616 77497-434
1-(3,5-Dichloro-2-fluorophenyl)-4,4-dimethylcyclohexanol ≥95%
1-(3,5-Dichloro-2-fluorophenyl)-4,4-dimethylcyclohexanol

Precaución: For research use only

Formula: C₁₄H₁₇Cl₂FO
MW: 291.189 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34777224
CAS Number: 2918851-83-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR