4-(2,3-Difluoro-4-methylphenyl)tetrahydro-2H-pyran-4-ol ≥95%

Supplier: AOBChem USA

Warning

102586-1G 102586-5G 102586-500MG
77463-804EA 597.4 USD
77463-804 77486-660 77476-612
4-(2,3-Difluoro-4-methylphenyl)tetrahydro-2H-pyran-4-ol ≥95%
4-(2,3-Difluoro-4-methylphenyl)tetrahydro-2H-pyran-4-ol

Precaución: For research use only

Formula: C₁₂H₁₄F₂O₂
MW: 228.235 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34777173
CAS Number: 1697638-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR