1-(2,3-Dichloropyridin-4-yl)cyclohexanol ≥97%

Supplier: AOBChem USA

Warning

102577-1G 102577-5G 102577-500MG 102577-25G
77463-800EA 415.09 USD
77463-800 77486-656 77476-608 77497-422
1-(2,3-Dichloropyridin-4-yl)cyclohexanol ≥97%
1-(2,3-Dichloropyridin-4-yl)cyclohexanol

Precaución: For research use only

Formula: C₁₁H₁₃Cl₂NO
MW: 246.133 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34777164
CAS Number: 2918888-29-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR