(5-Chlorothiophen-2-yl)trimethylsilane ≥97%

Supplier: AOBChem USA

Warning

102554-1G 102554-5G 102554-500MG
77463-796EA 648.9 USD
77463-796 77486-652 77476-604
(5-Chlorothiophen-2-yl)trimethylsilane ≥97%
(5-Chlorothiophen-2-yl)trimethylsilane

Precaución: For research use only

Formula: C₇H₁₁ClSSi
MW: 190.766 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06201822
CAS Number: 23395-56-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR