2-Bromo-5-(dimethoxymethyl)pyridine ≥97%
Supplier: AOBChem USA
|
Warning
|
102543-1G
102543-5G
102543-500MG
102543-25G
77463-792EA
258.53
USD
77463-792
77486-648
77476-600
77497-418
2-Bromo-5-(dimethoxymethyl)pyridine ≥97%
2-Bromo-5-(dimethoxymethyl)pyridine
Precaución: For research use only
Formula:
C₈H₁₀BrNO₂ MW: 232.075 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD25960047 CAS Number: 174608-37-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...