3-Chloro-2-ethoxy-4-(methylthio)pyridine ≥97%

Supplier: AOBChem USA

Warning

102411-1G 102411-5G 102411-500MG 102411-25G
77463-788EA 415.09 USD
77463-788 77486-644 77476-596 77497-410
3-Chloro-2-ethoxy-4-(methylthio)pyridine ≥97%
3-Chloro-2-ethoxy-4-(methylthio)pyridine

Precaución: For research use only

Formula: C₈H₁₀ClNOS
MW: 203.689 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34777031
CAS Number: 2901109-98-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR