2-(2-Bromo-6-fluoro-4-(trifluoromethyl)phenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

101870-1G 101870-5G 101870-500MG 101870-25G
77463-784EA 415.09 USD
77463-784 77486-640 77476-592 77497-386
2-(2-Bromo-6-fluoro-4-(trifluoromethyl)phenyl)-1,3-dioxolane ≥97%
2-(2-Bromo-6-fluoro-4-(trifluoromethyl)phenyl)-1,3-dioxolane

Precaución: For research use only

Formula: C₁₀H₇BrF₄O₂
Storage Temperature: Ambient
MDL Number: MFCD34776751
CAS Number: 2901105-74-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR