(4-(tert-Butyl)-2-(methoxymethoxy)phenyl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

101295-1G 101295-5G 101295-500MG
77463-776EA 555.17 USD
77463-776 77486-632 77476-584
(4-(tert-Butyl)-2-(methoxymethoxy)phenyl)boronic acid ≥95%
(4-(tert-Butyl)-2-(methoxymethoxy)phenyl)boronic acid

Precaución: For research use only

Formula: C₁₂H₁₉BO₄
Storage Temperature: Ambient
MDL Number: MFCD34622630
CAS Number: 161399-09-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR