2-Iodo-7-methoxynaphthalene ≥95%

Supplier: AOBChem USA

Warning

100981-1G 100981-5G 100981-500MG
77463-772EA 1269.99 USD
77463-772 77486-628 77476-580
2-Iodo-7-methoxynaphthalene ≥95%
2-Iodo-7-methoxynaphthalene

Precaución: For research use only

Formula: C₁₁H₉IO
Storage Temperature: Ambient
MDL Number: MFCD18412655
CAS Number: 128542-48-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR