7-Methoxy-2-naphthoic acid ≥95%

Supplier: AOBChem USA

Warning

100975-1G 100975-5G 100975-500MG
77463-768EA 785.89 USD
77463-768 77486-624 77476-576
7-Methoxy-2-naphthoic acid ≥95%
7-Methoxy-2-naphthoic acid

Precaución: For research use only

Formula: C₁₂H₁₀O₃
MW: 202.206 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18412886
CAS Number: 5043-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR