2-Fluoro-5-(methoxymethoxy)-3-propoxypyridine ≥95%

Supplier: AOBChem USA

Warning

100622-1G 100622-5G 100622-500MG
77463-764EA 451.14 USD
77463-764 77486-620 77476-572
2-Fluoro-5-(methoxymethoxy)-3-propoxypyridine ≥95%
2-Fluoro-5-(methoxymethoxy)-3-propoxypyridine

Precaución: For research use only

Formula: C₁₀H₁₄FNO₃
MW: 215.221 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34776401
CAS Number: 2918834-63-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR