2-(2-Fluoro-3-(trifluoromethyl)phenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

100406-1G 100406-5G 100406-500MG 100406-25G
77463-760EA 378.01 USD
77463-760 77486-616 77476-568 77497-328
2-(2-Fluoro-3-(trifluoromethyl)phenyl)propan-2-ol ≥95%
2-(2-Fluoro-3-(trifluoromethyl)phenyl)propan-2-ol

Precaución: For research use only

Formula: C₁₀H₁₀F₄O
MW: 222.179 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28357412
CAS Number: 1510239-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR