Isopropyl 3-(4-ethylphenyl)-3-hydroxypropanoate ≥95%

Supplier: AOBChem USA

343073-1G 343073-500MG 343073-5G 343073-25G
77469-874EA 415.09 USD
77469-874 77482-542 77492-964 77502-782
Isopropyl 3-(4-ethylphenyl)-3-hydroxypropanoate ≥95%
Isopropyl 3-(4-ethylphenyl)-3-hydroxypropanoate

Precaución: For research use only

Formula: C₁₄H₂₀O₃
CAS Number: 3002506-17-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR