Isopropyl 3-cyclopropyl-3-hydroxypropanoate ≥95%

Supplier: AOBChem USA

343067-1G 343067-500MG 343067-5G 343067-25G
77469-862EA 415.09 USD
77469-862 77482-530 77492-952 77502-770
Isopropyl 3-cyclopropyl-3-hydroxypropanoate ≥95%
Isopropyl 3-cyclopropyl-3-hydroxypropanoate

Precaución: For research use only

Formula: C₉H₁₆O₃
CAS Number: 3002460-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR