3-Bromo-2-fluoro-5-methylbenzyl alcohol ≥97%

Supplier: AOBChem USA

34298-1G 34298-500MG 34298-5G 34298-25G
77469-846EA 219.39 USD
77469-846 77482-514 77492-936 77502-760
3-Bromo-2-fluoro-5-methylbenzyl alcohol ≥97%
3-Bromo-2-fluoro-5-methylbenzyl alcohol

Precaución: For research use only

Formula: C₈H₈BrFO
MW: 219.051 g/mol
MDL Number: MFCD11110180
CAS Number: 307975-05-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR