1-(2-Bromo-6-fluoro-4-methylphenyl)-2,2,3,3,3-pentafluoropropan-1-one ≥95%

Supplier: AOBChem USA

342646-1G 342646-500MG 342646-5G 342646-25G
77469-826EA 415.09 USD
77469-826 77482-494 77492-916 77502-740
1-(2-Bromo-6-fluoro-4-methylphenyl)-2,2,3,3,3-pentafluoropropan-1-one ≥95%
1-(2-Bromo-6-fluoro-4-methylphenyl)-2,2,3,3,3-pentafluoropropan-1-one

Precaución: For research use only

Formula: C₁₀H₅BrF₆O
CAS Number: 3002520-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR