1-Bromo-4-fluoro-2-methoxy-3-methylbenzene ≥97%

Supplier: AOBChem USA

34264-1G 34264-500MG 34264-5G
77469-822EA 544.87 USD
77469-822 77482-492 77492-914
1-Bromo-4-fluoro-2-methoxy-3-methylbenzene ≥97%
1-Bromo-4-fluoro-2-methoxy-3-methylbenzene

Precaución: For research use only

Formula: C₈H₈BrFO
MDL Number: MFCD22414769
CAS Number: 1783385-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR