1-(4'-ethyl-[1,1'-biphenyl]-4-yl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

342229-1G 342229-500MG 342229-5G 342229-25G
77469-818EA 415.09 USD
77469-818 77482-486 77492-908 77502-736
1-(4'-ethyl-[1,1'-biphenyl]-4-yl)-2,2-difluoroethanone ≥95%
1-(4'-ethyl-[1,1'-biphenyl]-4-yl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₁₆H₁₄F₂O
CAS Number: 3002460-20-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR