Ethyl(4'-Ethyl-[1,1'-biphenyl]-4-yl)sulfane ≥95%

Supplier: AOBChem USA

342221-1G 342221-500MG 342221-5G 342221-25G
77469-814EA 415.09 USD
77469-814 77482-482 77492-904 77502-732
Ethyl(4'-Ethyl-[1,1'-biphenyl]-4-yl)sulfane ≥95%
Ethyl(4'-Ethyl-[1,1'-biphenyl]-4-yl)sulfane

Precaución: For research use only

Formula: C₁₆H₁₈S
CAS Number: 3002505-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR