1-(4'-ethyl-[1,1'-biphenyl]-4-yl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

342215-1G 342215-500MG 342215-5G 342215-25G
77469-810EA 415.09 USD
77469-810 77482-478 77492-900 77502-728
1-(4'-ethyl-[1,1'-biphenyl]-4-yl)-2,2,2-trifluoroethanone ≥95%
1-(4'-ethyl-[1,1'-biphenyl]-4-yl)-2,2,2-trifluoroethanone

Precaución: For research use only

Formula: C₁₆H₁₃F₃O
CAS Number: 3002448-59-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR