3-Butyl-5-formylbenzonitrile ≥95%

Supplier: AOBChem USA

342015-1G 342015-500MG 342015-5G 342015-25G
77469-806EA 415.09 USD
77469-806 77482-474 77492-896 77502-724
3-Butyl-5-formylbenzonitrile ≥95%
3-Butyl-5-formylbenzonitrile

Precaución: For research use only

Formula: C₁₂H₁₃NO
CAS Number: 3002542-33-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR