Cyclopropyl(2-fluoro-3-(trifluoromethyl)phenyl)methanol ≥95%

Supplier: AOBChem USA

341883-1G 341883-500MG 341883-5G
77469-790EA 646.84 USD
77469-790 77482-458 77492-880
Cyclopropyl(2-fluoro-3-(trifluoromethyl)phenyl)methanol ≥95%
Cyclopropyl(2-fluoro-3-(trifluoromethyl)phenyl)methanol

Precaución: For research use only

Formula: C₁₁H₁₀F₄O
CAS Number: 1536815-74-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR