1-Bromo-3-chloro-2-ethoxy-4-fluorobenzene ≥95%

Supplier: AOBChem USA

34028-1G 34028-500MG 34028-5G 34028-25G
77469-758EA 356.38 USD
77469-758 77482-428 77492-848 77502-684
1-Bromo-3-chloro-2-ethoxy-4-fluorobenzene ≥95%
1-Bromo-3-chloro-2-ethoxy-4-fluorobenzene

Precaución: For research use only

Formula: C₈H₇BrClFO
MDL Number: MFCD31619455
CAS Number: 2179038-46-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR