(2,3-Difluoro-4-(trifluoromethyl)phenyl)(4-methoxyphenyl)methanol ≥95%

Supplier: AOBChem USA

340164-1G 340164-500MG 340164-5G 340164-25G
77469-754EA 415.09 USD
77469-754 77482-424 77492-844 77502-682
(2,3-Difluoro-4-(trifluoromethyl)phenyl)(4-methoxyphenyl)methanol ≥95%
(2,3-Difluoro-4-(trifluoromethyl)phenyl)(4-methoxyphenyl)methanol

Precaución: For research use only

Formula: C₁₅H₁₁F₅O₂
CAS Number: 3002460-76-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR