1-(5-Bromo-2-fluoro-3-methoxyphenyl)propan-1-one ≥95%

Supplier: AOBChem USA

340151-1G 340151-500MG 340151-5G 340151-25G
77469-742EA 415.09 USD
77469-742 77482-412 77492-832 77502-670
1-(5-Bromo-2-fluoro-3-methoxyphenyl)propan-1-one ≥95%
1-(5-Bromo-2-fluoro-3-methoxyphenyl)propan-1-one

Precaución: For research use only

Formula: C₁₀H₁₀BrFO₂
CAS Number: 3002525-40-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR