1-(5-Bromo-2-fluoro-3-methoxyphenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

340147-1G 340147-500MG 340147-5G 340147-25G
77469-738EA 415.09 USD
77469-738 77482-408 77492-828 77502-666
1-(5-Bromo-2-fluoro-3-methoxyphenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(5-Bromo-2-fluoro-3-methoxyphenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₂H₁₆BrFO₂
MW: 291.1566 g/mol
CAS Number: 3002453-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR