1-(2,3-Difluoro-4-(trifluoromethyl)phenyl)-2-methylpropan-1-ol ≥95%

Supplier: AOBChem USA

340145-1G 340145-500MG 340145-5G 340145-25G
77469-734EA 415.09 USD
77469-734 77482-404 77492-824 77502-662
1-(2,3-Difluoro-4-(trifluoromethyl)phenyl)-2-methylpropan-1-ol ≥95%
1-(2,3-Difluoro-4-(trifluoromethyl)phenyl)-2-methylpropan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₁F₅O
CAS Number: 3002488-43-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR